Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
American Educational Products Astrolabe Kit
Perfect for introductory classes in astronomy!
Specifications
Product Type | Astrolabe kit |
---|---|
For Use With (Application) | Plotting, Locating movements of stars |
Grade | 7 to 12 |
Includes | Instructions |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
S53032
![]() |
American Educational Products
3014 |
1 kit |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
Description
- Measures declination, altitude, and azimuth of planets, stars, moon, and locations on the earth
- Plots and locates movements of stars May also be used as an inclinometer to measure angles of slope, strike, and dip, and for numerous other measurements
Specifications
Astrolabe kit | |
Plotting, Locating movements of stars | |
7 to 12 | |
Instructions |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title