Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASUN Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$436.00

Specifications

Antigen ASUN
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP170427
View Documents
Novus Biologicals
NBP170427
100 μL
Each of 1 for $436.00
Add to cart
 
Description

Description

ASUN Polyclonal specifically detects ASUN in Human samples. It is validated for Western Blot.
Specifications

Specifications

ASUN
Polyclonal
Rabbit
PBS and 2% Sucrose with 0.09% Sodium Azide
55726
Synthetic peptides corresponding to C12ORF11 The peptide sequence was selected from the middle region of C12orf11. Peptide sequence VQQHFDLASTTITNIPMKEEQHANTSANYDVELLHHKDAHVDFLKSGDSH.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
RUO
ASUN, asunder, spermatogenesis regulator homolog (Drosphila), cell cycle regulator Mat89Bb homolog, chromosome 12 open reading frame 11, FLJ10630, Mat89Bb, NET48, SPATA30
ASUN
IgG
Affinity Purified
80 kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit