Learn More
TWD Scientific LLC ASWF4XL 10/PK 6PK/CSASWF4XL 1
SDP
Supplier:  TWD Scientific LLC ASWF4XL
Encompass_Preferred
Disposable Anti-static Polypropylene Weighing Funnel. Extra Large. Fits Into 1000-2000ml Volumetric Flasks. 10/pack, Price/pack
Catalog No. NC0071147
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »