Learn More
Tocris Bioscience™ AT 101 Inhibitor
Low molecular weight, organic compound that may help regulate biological process.
Specifications
Description | CAS: 90141-22-3 |
---|---|
Formulation | C30H30O8 |
For Use With (Application) | Inhibits apoptosis |
Molecular Weight (g/mol) | 518.56g/mol |
Solubility | Soluble to 100mM in DMSO, 100mM in ethanol |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB336710MG
|
Tocris Bioscience™
NB336710MG |
10 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
NB336750MG
|
Tocris Bioscience™
NB336750MG |
50 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Biological Activity:
R-(-)-enantiomer of gossypol. Mimics BH3 domains of Bcl-2, Bcl-XL and Mcl-1. Disrupts heterodimerization of Bcl-2 with proapoptotic family members. Induces apoptosis in vitro through activation of caspase-9; cytotoxic to multiple myeloma and drug-resistant cell lines. Delays onset of androgen-independent growth of VCaP prostate cancer xenografts in vivo.
Storage: Store the unopened product at 2° to 8°C. Do not use past expiration date.
Specifications
CAS: 90141-22-3 | |
Inhibits apoptosis | |
Soluble to 100mM in DMSO, 100mM in ethanol | |
AT 101 |
C30H30O8 | |
518.56g/mol | |
-20°C | |
Dark Purple |
For Laboratory Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.