Learn More
Selleck Chemical LLCSupplier Diversity Partner AT9283 10mg 896466-04-9  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S113410MG
AT9283 is a potent JAK2/3 inhibitor with IC50 of 1.2 nM/1.1 nM in cell-free assays; also potent to Aurora A/B, Abl1(T315I). Phase 2.
Catalog No. 50-622-9