Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATAD2 Rabbit anti-Human, Human, Clone: ARC2317, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  Thermo Scientific MA538002

Encompass

Catalog No. PIMA538002


Add to cart

Description

Description

Positive Samples: MCF7, K-562

ATAD2 Recombinant Monoclonal antibody specifically detects ATAD2 in Human, Human samples. It is validated for Western Blot
Specifications

Specifications

ATAD2
Recombinant Monoclonal
0.84 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q6PL18
2610509G12Rik; AAA nuclear coregulator cancer-associated protein; ANCCA; Atad2; ATPase family AAA domain-containing protein 2; ATPase family, AAA domain containing 2; CT137; L16; PRO2000
A synthesized peptide derived from human ATAD2
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
ARC2317
Unconjugated
ATAD2
Liquid
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29028
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit