Learn More
ATAD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 166101AP
Catalog No. 50-173-1052
Add to cart
Description
Description
ATAD3B is a member of the AAA ATPase family, a family of proteins that catalyze ATP into ADP and are involved in several cellular functions such as cell-cycle regulation, protein proteolysis and transport. The AAA ATPase family is characterized by a highly conserved AAA motif containing Walker homology sequences and imparting ATPase activity. Mitochondrial membrane proteins ATAD3A/B contribute to the stabilization of nucleoids and may participate in the transformation pathway and the chemosensitivity of oligodendrogliomas. The gene encoding ATAD3A/B/C maps to human chromosome 1 and has been suggested to be an anti-apoptotic marker.
Specifications
Specifications
ATAD3
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5T9A4
AAA TOB3, ATAD3B, KIAA1273, TOB3
Rabbit
IgG
150 μL
-20°C
Primary
108888, 298682, 83858
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ATAD3B
Liquid
Atad3a, ATAD3B
ATAD3 Fusion Protein Ag9836
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat