Learn More
Cole-Parmer ATAGO 3442 PR-101A PALETTE-STY
Supplier:  Cole-Parmer 0294049
Encompass_Preferred
Atago 3442 PR-101A Palette-style digital refractometer, 0.0 to 45.0% Brix
Catalog No. NC2261585
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.