Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pocket Brix-Acidity Meter, PAL-BX/ACID

The dual-scale Atago™ Pocket Brix-Acidity Meter (PAL-BX/ACID) has revolutionized acid measurements. Simply place your sample on the prism and press the START button to read Brix, then dilute your sample with water and press START again to read the percent acid. 

Manufacturer:  Atago USA IncSupplier Diversity Partner 7104

 View more versions of this product

Catalog No. 05-100-250


  Check Availability
Add to cart

Specifications

Description

  • Easy-to-use compact, digital hand-held unit
  • Hybrid Brix and Acidity meter
  • Portable and quick alternative to titration, requires no costly reagent
  • Automatic Temperature Compensation (ATC)
  • Three second measurement time
  • Water-resistant
  • Stainless steel sample stage
  • LCD display
  • Backlight stays on for 30 seconds after any button is pressed
Specifications:
  • Brix Range: 0.0 to 60.0%
  • Brix Accuracy: ±0.2%
  • Brix Resolution: 0.1%
  • Acidity Range: 0.10 to 4.00%
  • Acidity Accuracy: ±0.10% (0.10 to 1.00%); Relative Precision ±10% (1.01 to 4.00%)
  • Acidity Resolution: 0.01%
  • ATC Range: 10° to 40°C
  • IP Rating: IP65
  • Sample Vol.: 0.2mL
  • Power Supply: two AAA Batteries
  • Dimensions (D x W x H): 31 x 55 x 109mm
  • Weight: 100g
Applications:
  • Cat. No. 05-100-267: For measuring acidity and brix of citrus fruits
  • Cat. No. 05-100-268: For measuring acidity and brix of Grapes and Wine
  • Cat. No. 05-100-269: For measuring acidity and brix of Tomatoes
  • Cat. No. 05-100-250: For measuring acidity and brix of Strawberry
  • Cat. No. 05-100-251: For measuring acidity and brix of Apple
  • Cat. No. 05-100-252: For measuring acidity and brix of Banana
  • Cat. No. 05-100-253: For measuring acidity and brix of Blueberry
  • Cat. No. 05-100-254: For measuring acidity and brix of Kiwi
  • Cat. No. 05-100-255: For measuring acidity and brix of Pineapple
  • Cat. No. 05-100-256: For measuring acidity and brix of Coffee Cherry
  • Cat. No. 05-100-257: For measuring acidity and brix of Milk
  • Cat. No. 05-100-258: For measuring acidity and brix of Yogurt
  • Cat. No. 05-100-259: For measuring acidity and brix of Beer
  • Cat. No. 05-100-260: For measuring acidity and brix of Sake
  • Cat. No. 05-100-261: For measuring acidity and brix of Vinegar
Description & Specifications

Specifications

Pocket Brix-Acidity Meter
LCD
Scale, two plastic beakers, an attachment, and five plastic pipets.
10° to 40°C
31 x 55 x 109mm
10° to 40°C
3 sec.
0.1°C
PAL-BX/ACID
Brix, citric and malic acid concentrations
Brix: ±0.2%, Acidity: ±0.10% (0.10 to 1.00%); Relative Precision ±10% (1.01 to 4.00%)
For measuring acidity and brix of Strawberry
Brix: 0.1%; Acidity: 0.01%
100g
±1°C
0.0 to 60.0%
Two AAA Batteries
0.2mL
IP65
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit