Pocket Brix-Acidity Meter, PAL-BX/ACID

The dual-scale Atago™ Pocket Brix-Acidity Meter (PAL-BX/ACID) has revolutionized acid measurements. Simply place your sample on the prism and press the START button to read Brix, then dilute your sample with water and press START again to read the percent acid. 

Manufacturer: Atago USA IncSupplier Diversity Partner 7105

 View more versions of this product

Catalog No. 05-100-251

  Check Availability
Add to Cart

    Specifications

    Includes Scale, two plastic beakers, an attachment, and five plastic pipets.
    Resolution Brix: 0.1%; Acidity: 0.01%
    Temperature Compensation 10° to 40°C
    Weight (Metric) 100g
    Dimensions (L x W x H) 31 x 55 x 109mm
    Accuracy (Temperature) ±1°C
    Ambient Temperature (Metric) 10° to 40°C
    Brix Range 0.0 to 60.0%
    Measurement Time 3 sec.
    Power Supply Details Two AAA Batteries
    Temperature Resolution 0.1°C
    Sample Volume 0.2mL
    Model PAL-BX/ACID
    IP Rating IP65
    Scale Type Brix, malic acid concentrations
    Accuracy Brix: ±0.2%, Acidity: ±0.10% (0.10 to 1.00%); Relative Precision ±10% (1.01 to 4.00%)
    Display LCD
    For Use With (Application) For measuring acidity and brix of Apple
    Product Type Pocket Brix-Acidity Meter

    • Easy-to-use compact, digital hand-held unit
    • Hybrid Brix and Acidity meter
    • Portable and quick alternative to titration, requires no costly reagent
    • Automatic Temperature Compensation (ATC)
    • Three second measurement time
    • Water-resistant
    • Stainless steel sample stage
    • LCD display
    • Backlight stays on for 30 seconds after any button is pressed
    Specifications:
    • Brix Range: 0.0 to 60.0%
    • Brix Accuracy: ±0.2%
    • Brix Resolution: 0.1%
    • Acidity Range: 0.10 to 4.00%
    • Acidity Accuracy: ±0.10% (0.10 to 1.00%); Relative Precision ±10% (1.01 to 4.00%)
    • Acidity Resolution: 0.01%
    • ATC Range: 10° to 40°C
    • IP Rating: IP65
    • Sample Vol.: 0.2mL
    • Power Supply: two AAA Batteries
    • Dimensions (D x W x H): 31 x 55 x 109mm
    • Weight: 100g
    Applications:
    • Cat. No. 05-100-267: For measuring acidity and brix of citrus fruits
    • Cat. No. 05-100-268: For measuring acidity and brix of Grapes and Wine
    • Cat. No. 05-100-269: For measuring acidity and brix of Tomatoes
    • Cat. No. 05-100-250: For measuring acidity and brix of Strawberry
    • Cat. No. 05-100-251: For measuring acidity and brix of Apple
    • Cat. No. 05-100-252: For measuring acidity and brix of Banana
    • Cat. No. 05-100-253: For measuring acidity and brix of Blueberry
    • Cat. No. 05-100-254: For measuring acidity and brix of Kiwi
    • Cat. No. 05-100-255: For measuring acidity and brix of Pineapple
    • Cat. No. 05-100-256: For measuring acidity and brix of Coffee Cherry
    • Cat. No. 05-100-257: For measuring acidity and brix of Milk
    • Cat. No. 05-100-258: For measuring acidity and brix of Yogurt
    • Cat. No. 05-100-259: For measuring acidity and brix of Beer
    • Cat. No. 05-100-260: For measuring acidity and brix of Sake
    • Cat. No. 05-100-261: For measuring acidity and brix of Vinegar

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit