Learn More
Selleck Chemical LLCSupplier Diversity Partner Ataluren (PTC124) 50mg 775304-57-9  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S600350MG
Ataluren (PTC124) selectively induces ribosomal read-through of premature but not normal termination codons, with EC50 of 0.1 μM in HEK293 cells, may provide treatment for genetic disorders caused by nonsense mutations (e.g. CF caused by CFTR nonsense mutation). Phase 3.
Catalog No. 50-699-5