Learn More
Ataxin 1 Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: S76-8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242186UV
Catalog No. NBP242186UV
Add to cart
Description
Description
Ataxin 1 Monoclonal antibody specifically detects Ataxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
Ataxin 1
S76-8
Western Blot, Immunohistochemistry
Purified
ATXN1
Synthetic peptide amino acids 164-197 (ATTPSQRSQLEAYSTLLANMGSLSQAPGHKVEPP) of mouse Ataxin-1. Rat: 100% identity (34/34 amino acids identical). Human: 88% identity (30/34 amino acids identical).
Protein G purified
Phospho Specific
Primary
6310
Human, Mouse, Rat
Western Blot, Immunohistochemistry
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
ataxin 1, ataxin 1), ataxin-1, ATX1spinocerebellar ataxia 1 (olivopontocerebellar ataxia 1, autosomal dominant, SCA1D6S504E, Spinocerebellar ataxia type 1 protein
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 85kDa.