Learn More
Ataxin 1 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: S65-37, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242204FR
Catalog No. NBP242204FR
Add to cart
Description
Description
Ataxin 1 Monoclonal antibody specifically detects Ataxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
Ataxin 1
S65-37
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
ATXN1
Synthetic peptide amino acids 746-761 (RKRRWSAPETRKLEKS) of mouse ataxin-1. Rat: 93% identity (15/16 amino acids identical). Human: 87% identity (14/16 amino acids identical).
Protein G purified
Phospho Specific
Primary
6310
Human, Mouse, Rat
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
ataxin 1, ataxin 1), ataxin-1, ATX1spinocerebellar ataxia 1 (olivopontocerebellar ataxia 1, autosomal dominant, SCA1D6S504E, Spinocerebellar ataxia type 1 protein
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 85kDa. No cross-reactivity against phosphor S751-Ataxin-1.