Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ataxin 7 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31046925UL

 View more versions of this product

Catalog No. NB125840


Add to cart

Description

Description

Ataxin 7 Polyclonal antibody specifically detects Ataxin 7 in Rat samples. It is validated for Western Blot
Specifications

Specifications

Ataxin 7
Unconjugated
PBS buffer, 2% sucrose
ADCAII, ataxin 7, FLJ17787, SCA7
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat Ataxin 7 (XP_001069337). Peptide sequence MSERAADDVRGEPRRAAGGAAAARQQQQPQPLQLQRQHPPPRRPRPEDGG
25 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Rat
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Primary
6314
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit