ATC 0065, Tocris

Potent and selective MCH1 antagonist

Manufacturer: R&D Systems 3359/10

Quantity 10mg

 View more versions of this product

Catalog No. 33-591-0

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C25H29BrF3N5O·2HCl
    Formula Weight 625.35
    Synonym N2-[cis-4-[[2-[4-Bromo-2-(trifluoromethoxy)phenyl]ethyl]amino]cyclohexyl]-N4, N4-dimethyl-2, 4-quinazolinediamine dihydrochloride
    Chemical Name or Material ATC 0065

    Specifications

    Formula Weight 625.35
    Quantity 10mg
    Assay Percent Range >98%
    Molecular Formula C25H29BrF3N5O·2HCl
    CAS 510732-84-0
    Synonym N2-[cis-4-[[2-[4-Bromo-2-(trifluoromethoxy)phenyl]ethyl]amino]cyclohexyl]-N4, N4-dimethyl-2, 4-quinazolinediamine dihydrochloride
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material ATC 0065

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit