Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atenolol, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
29122-68-7
C14H22N2O3
266.34
METKIMKYRPQLGS-UHFFFAOYNA-N
2-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)acetamide
CC(C)NCC(O)COC1=CC=C(CC(N)=O)C=C1
29122-68-7
C14H22N2O3
266.34
METKIMKYRPQLGS-UHFFFAOYNA-N
2-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)acetamide
CC(C)NCC(O)COC1=CC=C(CC(N)=O)C=C1

A1648, 29122-68-7, C14H22N2O3

Spectrum™ Atenolol, USP A receptor antagonist that belongs to the beta blocker group. It is a drug used to treat cardiovascular diseases by slowing the heart beat down and in turn reducing the strain on the heart. It is used for several conditions that include supraventricular tachycardia, hypertension, angina and acute myocardial infarction. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.005
Ignition Residue 0.002
Melting Range 152-156.5°C
Quantity 1 g
CAS Max % 1
Percent Purity 98 to 102%