Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Atenolol, USP, 98-102%, Spectrum™
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Description
Specifications
Ignition Residue | 0.2% |
Loss on Drying | 0.5% |
Packaging | Amber Glass Bottle |
Melting Range | 152-156.5°C |
Quantity | 5 g |
CAS Max % | 100% |
Percent Purity | 98 to 102% |
Grade | USP |