Learn More
ATF1 Antibody (OTI2B4), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270218UV
Catalog No. NBP270218UV
Add to Cart
Description
Description
ATF1 Monoclonal specifically detects ATF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
ATF1
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
ATF1
Full length recombinant protein of human ATF1 (NP_005162) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2b
Western Blot, Immunohistochemistry
OTI2B4
Western Blot, Immunohistochemistry
activating transcription factor 1FUS/ATF-1, cAMP-dependent transcription factor ATF-1, EWS-ATF1, Protein TREB36, TREB36cyclic AMP-dependent transcription factor ATF-1
Mouse
Affinity Purified
Phospho Specific
466.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions