Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
ATF2 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer: Abnova Corporation H00001386T01
Description
Western Blotting
Specifications
293T | |
BC026175 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-ATF2 antibody (H00001386-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Human |
ATF2 293T Cell Transient Overexpression Lysate(Denatured) | |
23.1 | |
pCMV-ATF2 full-length | |
100 uL | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Western Blot |