Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF2 Mouse anti-Human, Clone: PCRP-ATF2-1B4, Novus Biologicals™
Mouse Monoclonal Antibody
$208.09 - $439.77
Specifications
Antigen | ATF2 |
---|---|
Clone | PCRP-ATF2-1B4 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array |
Description
ATF2 Monoclonal antibody specifically detects ATF2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
ATF2 | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array | |
Unconjugated | |
Mouse | |
Cancer, Chromatin Research, Immunology, Innate Immunity, Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558 | |
Recombinant full-length human ATF2 protein (Uniprot: P15336) | |
Primary | |
Store at 4°C. |
PCRP-ATF2-1B4 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P15336 | |
1386 | |
IgG2b | |
Protein A or G purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title