Learn More
ATF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148341AP
Catalog No. 50-172-9764
Add to cart
Description
Description
This antibody is a rabbit polyclonal antibody raised against a peptide mapping within human ATF2. The calculated molecular weight of ATF2 is 55 kDa, but the modified ATF2 protein is about 70 kDa.
The transcription factor ATF-2 (also called CRE-BP1) binds to both AP-1 and CRE DNA response elements and is a member of the ATF/CREB family of leucine zipper proteins. ATF-2 interacts with a variety of viral oncoproteins and cellular tumor suppressors and is a target of the SAPK/JNK and p38 MAP kinase signaling pathways. Various forms of cellular stress, including genotoxic agents, inflammatory cytokines and UV irradiation, stimulate the transcriptional activity of ATF-2. Cellular stress activates ATF-2 by phosphorylation of Thr69 and Thr71. Both SAPK and p38 MAPK have been shown to phosphorylate ATF-2 at these sites in vitro and in cells transfected with ATF-2.
Specifications
Specifications
ATF2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15336, P16951, Q00969
ATF2, CRE BP1, CREB 2, CREB2, CREBP1, HB16, TREB7
Rabbit
IgG
150 μL
-20°C
Primary
11909, 1386, 81647
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ATF2
Liquid
Atf2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat