Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF5 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31065225UL

 View more versions of this product

Catalog No. NB126206


Add to cart

Description

Description

ATF5 Polyclonal antibody specifically detects ATF5 in Rat samples. It is validated for Western Blot
Specifications

Specifications

ATF5
Unconjugated
PBS buffer, 2% sucrose
activating transcription factor 5FLJ34666, ATFXHMFN0395, cAMP-dependent transcription factor ATF-5, cyclic AMP-dependent transcription factor ATF-5, Transcription factor ATFx
The immunogen is a synthetic peptide directed towards the C-terminal region of Rat ATF5 (NP_758839). Peptide sequence EGEALEGECQGLEARNRELRERAESVEREIQYVKDLLIEVYKARSQRTRS
25 μg
Cell Cycle and Replication
Primary
22809
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit