Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG12 Mouse anti-Human, Mouse, DyLight 680, Clone: 9411, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243753FR

Catalog No. NBP243753FR


Add to cart

Description

Description

ATG12 Monoclonal antibody specifically detects ATG12 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

ATG12
9411
Western Blot
Purified
ATG12
The exact sequence is proprietary.
Protein G purified
Apoptosis, Autophagy, Neuroscience
Primary
9140
Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Apg12 (autophagy 12, S. cerevisiae)-like, APG12 autophagy 12-like (S. cerevisiae), APG12HAPG12, APG12-like, ATG12 autophagy related 12 homolog (S. cerevisiae), Autophagy-related protein 12, FBR93, ubiquitin-like protein ATG12, yeast) homolog
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit