Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG16L1 Rabbit anti-Human, Mouse, Rat, Bovine, Canine, Primate, DyLight™ 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11060928UV

Catalog No. N11060928UV


Add to cart

Description

Description

ATG16L1 Polyclonal antibody specifically detects ATG16L1 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

ATG16L1
DyLight 350
50 mM sodium borate with 0.05% sodium azide
APG16 autophagy 16-like (S. cerevisiae), APG16L beta, APG16LFLJ10828, APG16-like 1, ATG16 autophagy related 16-like (S. cerevisiae), ATG16 autophagy related 16-like 1 (S. cerevisiae), ATG16A, ATG16L, autophagy-related protein 16-1, FLJ00045, FLJ10035, FLJ22677, IBD10, WD repeat domain 30, WDR30
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human, Mouse, Rat, Bovine, Canine, Primate
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
ATG16L1
A synthetic peptide within residues 1-100 of human ATG16L1 protein. [Swiss-ProtNo. Q676U5]
Immunogen affinity purified
Autophagy, Cancer, Cell Biology, Macroautophagy, Neurodegeneration, Neuroscience, Phospho Specific, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary
55054.0
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit