Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG2A Mouse anti-Human, Clone:4E6D4, Abnova™

Mouse monoclonal antibody raised against recombinant human ATG2A.

Supplier:  Abnova Corporation MAB17284

Catalog No. 89-333-515


Add to Cart

Description

Description

Specifications

Specifications

ATG2A
Monoclonal
Unconjugated
ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry (1:50- 1:200)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
ATG2A
Mouse
100 μg
Human
Antibody
IgG1
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
4E6D4
ATG2 autophagy related 2 homolog A (S. cerevisiae)
In PBS (0.05% sodium azide)
KIAA0404, MGC117153
Recombinant protein corresponding to amino acid 325-429 of human ATG2A from E. coli.
23130
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit