Learn More
ATG3 Mouse anti-Human, Clone: OTI2B9, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA503384
Catalog No. 50-167-4290
Add to cart
Description
Description
ATG3 (APG3) is a widely expressed conjugating enzyme for APG8 lipidation, an essential step for the initiation of autophagy. Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. Autophagy is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. ATG3 functions as an E2-like enzyme during the initial stages of autophagosome formation by catalyzing the formation of the Atg8-phosphatidylethanolamine (Atg8-PE) conjugate, which is critical for autophagy.
Specifications
Specifications
ATG3
Monoclonal
Unconjugated
ATG3
Liquid
ATG3
Full length human recombit protein of human ATG3 produced in HEK293 cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2B9
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NT62
APG3, APG3-LIKE, APG3L, PC3-96
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64422