Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG4B Polyclonal Antibody
ATG4B Polyclonal Antibody
Supplier: Thermo Scientific PA5114497
Description
ATG4B Polyclonal Antibody for Western Blot, IHC (P)

Specifications
ATG4B | |
Polyclonal | |
Unconjugated | |
Atg4b | |
2510009N07Rik; APG4 (ATG4) autophagy-related homolog B; APG4 autophagy 4 homolog B; APG4B; ATG4 autophagy related 4 homolog B; Atg4b; atg4b protein; Atg4bl; AUT2B; Aut2B1; Aut2b2; AUTL1; AUT-like 1 cysteine; AUT-like 1 cysteine endopeptidase; AUT-like 1, cysteine endopeptidase; autophagin 1; autophagin-1; Autophagin-2B; autophagy related 4 homolog B; autophagy related 4B cysteine peptidase; autophagy related 4B, cysteine peptidase; autophagy-related 4B; autophagy-related cysteine endopeptidase 1; Autophagy-related cysteine endopeptidase 2B; autophagy-related protein 4 homolog B; AW048066; bAut2B; Cysteine protease ATG4B; cysteine protease involved in autophagy APG4-B; hAPG4B; KIAA0943 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23192, 316640, 66615 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8BGE6, Q9Y4P1 | |
Atg4b | |
ATG4B antibody was raised against a 17 amino acid peptide near the carboxy terminus of human ATG4B. The immunogen is located within the last 50 amino acids of ATG4B. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction