Learn More
ATG4C Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 203821AP
Catalog No. 50-173-2857
Add to cart
Description
Description
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some maligt tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Alternate transcriptional splice variants, encoding the same protein, have been characterized. Cysteine protease required for autophagy, which cleaves the C-terminal part of either MAP1LC3, GABARAPL2 or GABARAP, allowing the liberation of form I. A subpopulation of form I is subsequently converted to a smaller form (form II). Form II, with a revealed C-terminal glycine, is considered to be the phosphatidylethanolamine (PE)-conjugated form, and has the capacity for the binding to autophagosomes. Highly expressed in skeletal muscle, heart, liver and testis.
Specifications
Specifications
ATG4C
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q811C2, Q96DT6
APG4 C, APG4C, ATG4C, AUTL1, AUTL3, Autophagin 3, Cysteine protease ATG4C
Rabbit
IgG
150 μL
-20°C
Primary
242557, 84938
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ATG4C
Liquid
ATG4C
ATG4C Fusion Protein Ag14207
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse