Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG5 Rabbit anti-Human, Clone: AGT5/3220R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen ATG5
Clone AGT5/3220R
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123196
View Documents
Novus Biologicals
NBP30786120UG
20 μg
Each for $159.82
Add to cart
 
NB123195
View Documents
Novus Biologicals
NBP30786110UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

ATG5 Monoclonal antibody specifically detects ATG5 in Human samples. It is validated for ELISA
Specifications

Specifications

ATG5
0.2 mg/ml
ELISA
Purified
RUO
Human
APG5, APG5 (Autophagy 5, S. Cerevisiae)-Like, APG5 autophagy 5-like (S. cerevisiae), APG5L, APG5-LIKE, ASP, ASPAPG5-LIKE, ATG5 Autophagy Related 5 Homolog, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5, Autophagy Related 5, HAPG5, SCAR25
Recombinant fragment of human ATG5 protein (around aa 1-119) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
AGT5/3220R
ELISA
Unconjugated
Rabbit
Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway
10 mM PBS with 0.05% BSA
9474
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit