Learn More
ATG9A Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Supplier:  Thermo Scientific OSA00009G
Catalog No. PIOSA00009G
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 500 μL of distilled water. Centrifuge to remove any insoluble material.
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some maligt tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATG9A
Polyclonal
ATG9A
ATG9 autophagy related 9 homolog A (S. cerevisiae), ATG9A, APG9L1, APG9-like 1, MGD3208
Sheep
Ammonium sulfate precipitation
RUO
245860, 363254, 79065
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry, Western Blot
Unconjugated
Q7Z3C6, Q68FE2, Q5FWU3
ATG9A
A synthetic peptide from human autophagy-related protein 9A
500 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Recommendations
Recommendations