Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATH 5 Rabbit anti-Human,Mouse,Rat, KHL, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$153.50 - $395.00

Specifications

Antigen ATH 5
Applications Western Blot, Immunocytochemistry
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AB5694I10MI
View Documents
MilliporeSigma
AB5694I100UG
100 μg
Each for $395.00
Add to cart
 
AB5694I25MI
View Documents
MilliporeSigma
AB5694I25UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

ATH 5 Polyclonal antibody specifically detects ATH 5 in Human,Mouse,Rat samples. It is validated for Western Blotting, Immunocytochemistry.
Specifications

Specifications

ATH 5
KHL
Rabbit
Protein atonal homolog 7;Class A basic helix-loop-helix protein 13;bHLHa13;Helix-loop-helix protein hATH-5
KLH-conjugated linear peptide corresponding to 19 amino acids from within the bHLH domain.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Western Blot, Immunocytochemistry
Purified
Human, Mouse, Rat
ATOH7, ATH5, BHLHA13
Polyclonal
Affinity Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit