Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATIC Mouse anti-Canine, Human, Clone: OTI4A10, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504570

Catalog No. 50-167-5226


Add to cart

Description

Description

ATIC is a 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase which helps catalyze the penultimate and final steps of the de novo purine biosynthetic pathway. The human ATIC cDNA encodes a deduced 591-amino acid protein that is 81% identical to the chicken sequence.
Specifications

Specifications

ATIC
Monoclonal
3.51 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P31939
AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
471, 488513
Western Blot
OTI4A10
Unconjugated
ATIC
Liquid
ATIC
Full length human recombit protein of human ATIC produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit