Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATIC Mouse anti-Human, Mouse, Rat, Canine, Monkey, PE/Atto594, Clone: OTI1D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270220PEATT594

Catalog No. N270220P594


Add to cart

Description

Description

ATIC Monoclonal antibody specifically detects ATIC in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

ATIC
OTI1D2
Flow Cytometry
Affinity Purified, Conjugated
ATIC
Full length recombinant protein of human ATIC (NP_004035) produced in HEK293T cells.
Immunogen affinity purified
Stem Cell Markers
Primary
471.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMPcyclohydrolase, 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotidetransformylase/inosinicase, AICAR, AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme, AICARFT, AICARFT/IMPCHASE, bifunctional purine biosynthesis protein PURH, IMPCHASE, phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase, PURHFLJ93545
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit