Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotix™ aTIP Beckman™ Biomek™ Filter Pipette Tips Available on GSA/VA Contract for Federal Government customers only.

For use with Beckman Biomek FX/NX, AP384, 384-Head platforms.

$718.14 - $748.74

Specifications

Color Clear
Sterility Sterile
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Tip Style Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Tip Style Price Quantity & Availability  
12-111-065 Available on GSA/VA Contract for Federal Government customers only. View Documents Biotix™
BA00303FC
30μL Standard Length Pack of 10 for $718.14
Case of 5 PK for $3,590.00
Add to cart
 
12-111-067 Available on GSA/VA Contract for Federal Government customers only. View Documents Biotix™
BA00513FC
30μL Extended Length Pack of 10 for $748.74
Case of 5 PK for $3,743.00
Add to cart
 
Description

Description

  • Made from clear polypropylene
  • With graduation marks

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Clear
Sterile
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit