Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotix™ aTIP Non-Filter Pipette Tips
For use with Beckman Biomek FX/NX, Multimek, AP96 & Biomek 3000, 96-Head platforms.
Specifications
Filtered | Non-Filtered |
---|---|
Color | Clear |
Autoclavable | Autoclavable |
Format | Rack |
Material | Virgin Polypropylene |
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Price | Quantity & Availability | ||||
12-111-053
|
Biotix™
B00209NC |
20 μL | Non-sterile | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
12-111-057
|
Biotix™
BA02509SC |
250 μL | Sterile | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Made from medical grade virgin polypropylene
- Clear color
Specifications
Non-Filtered | |
Autoclavable | |
Virgin Polypropylene |
Clear | |
Rack |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title