Learn More
Waters Corp Waters Column, dC18, 3 μm, 2.1 mm x 30 mm, 100 Å
Supplier:  Waters Corp 186001287
Encompass_Preferred
This silica-based, reversed-phase C18 column is designed for polar compound retention in highly aqueous mobile phases, including 100% water. It provides balanced retention for polar and non-polar molecules, is LC/MS compatible, and offers improved pH stability. Optimized stationary phase properties ensure superior peak shape and resistance to dewetting.
  • Universal, silica-based, reversed-phase C18 column.
  • Optimized for polar compound retention in highly aqueous mobile phases.
  • LC/MS compatible with balanced retention for polar and non-polar molecules.
  • Superior peak shape, improved pH stability, and resistance to dewetting.
  • Specialized packing material for water-soluble chemicals.
  • Ensures prolonged column lifetime and superb peak shapes for bases.
Catalog No. 50-819-054