Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATM Antibody (2C1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100309
Description
ATM Monoclonal specifically detects ATM in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, SDS-Page, Single Cell Western, Knockdown Validated.Specifications
ATM | |
Monoclonal | |
1.0mg/mL | |
Western Blot 1:500 - 1:3000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1 - 10 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, SDS-Page, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Single Cell Western 100 ug/mL, Knockdown Validated | |
AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated (includes complementation groups A, C and D), ataxia telangiectasia mutatedATD, ATC, ATDC, ATE, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1 | |
Mouse | |
370 kDa | |
100 μL | |
p53 Pathway | |
472 | |
Human, Mouse, Rat, Monkey | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot | |
2C1 | |
Unconjugated | |
Q13315 | |
ATM | |
Recombinant protein expressed in E. coli corresponding to amino acids 2577-3056. | |
Antigen Affinity-purified | |
RUO | |
Primary | |
NB100-309 recognizes full-length ATM, a 350-kDa nuclear phosphoprotein. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction