Learn More
Selleck Chemical LLCSupplier Diversity Partner ATN-161 (Ac-PHSCN-NH2) DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S84545MG
ATN-161 (Ac-PHSCN-NH2) is a novel small peptide antagonist of integrin α5β1. It binds to several integrins, including α5β1 and αvβ3, that play a role in angiogenesis and tumor progression.
Catalog No. 50-194-8118