Learn More
ATN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 192431AP
Catalog No. 50-173-2373
Add to cart
Description
Description
Dentatorubral pallidoluysian atrophy (DRPLA) is a rare neurodegenerative disorder characterized by cerebellar ataxia, myoclonic epilepsy, choreoathetosis, and dementia. The disorder is related to the expansion from 7-35 copies to 49-93 copies of a trinucleotide repeat (CAG/CAA) within this gene. The encoded protein includes a serine repeat and a region of alternating acidic and basic amino acids, as well as the variable glutamine repeat. Alternative splicing results in two transcripts variants that encode the same protein.
Specifications
Specifications
ATN1
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35126, P54259
ATN1, atrophin 1, B37, D12S755E, DRPLA, HRS, NOD
Rabbit
IgG
150 μL
-20°C
Primary
13498, 1822
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ATN1
Liquid
ATN1
ATN1 Fusion Protein Ag5835
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse