Learn More
Atomic Models Set Products for Science Education
Visualize the structure of atoms and their respective components.
Manufacturer:  Denoyer Geppert Company FOM611
 View more versions of this product
Catalog No. S19433A
Add to cart
Description
Description
  • Set consists of 10 atoms showing electrons, protons, and neutrons
  • Atoms represented include: hydrogen, helium, lithium, beryllium, boron, carbon, nitrogen, oxygen, fluorine, and neon
  • Isotopes for the models of hydrogen, helium, and lithium can be made with a magnetically attached neutron
  • Optional stand is also available
Specifications
Specifications
Education
Hydrogen, Helium, Lithium, Beryllium, Boron, Carbon, Nitrogen, Oxygen, Fluorine and Neon
Stand For Atomic Models