Learn More
Atomic Models Set Products for Science Education
Visualize the structure of atoms and their respective components.
$119.25
Save up to 25%
Specifications
Includes Hydrogen, Helium, Lithium, Beryllium, Boron, Carbon, Nitrogen, Oxygen, Fluorine and Neon
View More Specs
Products 1
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
S194331 Products for Science Education
View Documents
Denoyer Geppert Company
FOM622
Atomic Model Set
Each for $119.25
Save 25% Reg : $159.01
Add to cart
Description
Description
  • Set consists of 10 atoms showing electrons, protons, and neutrons
  • Atoms represented include: hydrogen, helium, lithium, beryllium, boron, carbon, nitrogen, oxygen, fluorine, and neon
  • Isotopes for the models of hydrogen, helium, and lithium can be made with a magnetically attached neutron
  • Optional stand is also available
Specifications
Specifications
Hydrogen, Helium, Lithium, Beryllium, Boron, Carbon, Nitrogen, Oxygen, Fluorine and Neon