Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atovaquone, USP, 97.5-101.5%, Spectrum™

Click to view available options
Quantity:
500 g
1 kg
Packaging:
Amber Glass Bottle
95233-18-4
C22H19ClO3
366.84
BSJMWHQBCZFXBR-UHFFFAOYSA-N
3-[4-(4-chlorophenyl)cyclohexyl]-4-hydroxy-1,2-dihydronaphthalene-1,2-dione
OC1=C(C2CCC(CC2)C2=CC=C(Cl)C=C2)C(=O)C(=O)C2=CC=CC=C12
95233-18-4
C22H19ClO3
366.84
BSJMWHQBCZFXBR-UHFFFAOYSA-N
3-[4-(4-chlorophenyl)cyclohexyl]-4-hydroxy-1,2-dihydronaphthalene-1,2-dione
OC1=C(C2CCC(CC2)C2=CC=C(Cl)C=C2)C(=O)C(=O)C2=CC=CC=C12

A2154, 95233-18-4, C22H19ClO3

Spectrum™ Atovaquone, USP is an antibiotic sometimes prescribed to treat pneumocystis pneumonia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Quantity 500 g
CAS Max % 1
Percent Purity 97.5 to 101.5%