Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP Citrate Lyase Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI3G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270073JF549

Catalog No. N270073J549


Add to cart

Description

Description

ATP Citrate Lyase Monoclonal antibody specifically detects ATP Citrate Lyase in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

ATP Citrate Lyase
OTI3G8
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
ACLY
Full length recombinant protein of human ATP Citrate Lyase (NP_001087) produced in HEK293T cells.
Immunogen affinity purified
Phospho Specific
Primary
47.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
ACLEC 2.3.3.8, ATP citrate lyase, ATP citrate synthase, ATP-citrate (pro-S-)-lyase, ATP-citrate synthase, ATPCL, Citrate cleavage enzyme, CLATP
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit