Learn More
ATP Citrate Lyase Mouse anti-Human, Mouse, Rat, Monkey, DyLight 755, Clone: 5F8D11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237429IR
Catalog No. NBP237429IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ATP Citrate Lyase
5F8D11
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
ACLY
Purified recombinant fragment of human ACLY (AA: 306-502 ) expressed in E. Coli.
Protein A or G purified
Phospho Specific
Primary
47
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
ACLEC 2.3.3.8, ATP citrate lyase, ATP citrate synthase, ATP-citrate (pro-S-)-lyase, ATP-citrate synthase, ATPCL, Citrate cleavage enzyme, CLATP
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Monkey, Mouse, Rat