Learn More
ATP Synthase gamma Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA590325
Description
Immunogen sequence: ATLKDITRRL KSIKNIQKIT KSMKMVAAAK YARAERELKP ARIYGLGSLA LYEKADIKGP EDKKKHLLIG VSSDRGLCGA IHSSIAKQMK SEVATLTAAG KEVMLVGIGD KIRGILYRTH SDQFLVAFKE VGRKPPTFGD ASVIALELLN SGYEFDEGSI IFNKFRSVIS YKTEEKPIFS LNTVASADSM SIYDDIDADV LQNYQEYNLA NIIYYSLKES TTSEQSARMT AMDNASKNAS EMIDKLTLTF NRTRQAVITK ELIEIISGAA ALD; Positive Samples: LO2, U-87MG, Mouse brain, Mouse heart, Mouse kidney, Mouse liver, Rat brain, Rat liver; Cellular Location: Mitochondrion, Mitochondrion inner membrane, Peripheral membrane protein
ATP5C1 encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3.Specifications
ATP Synthase gamma | |
Polyclonal | |
Unconjugated | |
ATP5C1 | |
Atp5c1 | |
Recombit fusion protein containing a sequence corresponding to amino acids 26-298 of human ATP5C1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
2.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35435, P36542, Q91VR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116550, 11949, 509 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.