Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP10D Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen ATP10D
Immunogen Synthetic peptides corresponding to Atp10d (ATPase, class V, type 10D) The peptide sequence was selected from the middle region of Atp10d. Peptide sequence YAKRGLRTLCVAKKVMSDTEYAEWLRNHFLAETSIDNREELLVESAMRLE.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB16920020 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16911120UL
20ul Each for $149.00
Add to cart
 
NBP169111 View Documents Novus BiologicalsSupplier Diversity Partner
NBP169111
100 ul Each for $329.00
Add to cart
 
Description

Description

ATP10D Polyclonal antibody specifically detects Antigen in Rat samples. It is validated for Western Blotting.
Specifications

Specifications

ATP10D
Rabbit
Polyclonal
Unconjugated
57205
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to Atp10d (ATPase, class V, type 10D) The peptide sequence was selected from the middle region of Atp10d. Peptide sequence YAKRGLRTLCVAKKVMSDTEYAEWLRNHFLAETSIDNREELLVESAMRLE.
Primary
IgG
ATPase, class V, type 10D, ATPVDtype IV aminophospholipid transporting ATPase, EC 3.6.3, EC 3.6.3.1, KIAA1487ATPase class V type 10D, probable phospholipid-transporting ATPase VD
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit