Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP11A Upstream Neighbor Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen ATP11A Upstream Neighbor
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126418
View Documents
Novus Biologicals
NBP31075825UL
25 μg
Each for $106.19
Add to cart
 
NB126417
View Documents
Novus Biologicals
NBP310758100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

ATP11A Upstream Neighbor Polyclonal antibody specifically detects ATP11A Upstream Neighbor in Human samples. It is validated for Western Blot
Specifications

Specifications

ATP11A Upstream Neighbor
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
400165
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
C13orf35, C13orf35 chromosome 13 open reading frame 35
The immunogen is a synthetic peptide directed towards the C-terminal region of Human C13orf35 (NP_997323). Peptide sequence YKGRRSASGTQKQLQLPDTLSSLLCWRGAIMVYIKVTVQTDDSNKLLSLL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit