Learn More
ATP13A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 162441AP
Catalog No. 50-173-0823
Add to cart
Specifications
Specifications
ATP13A1
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9EPE9, Q9HD20
ATP13A, ATP13A1, ATPase type 13A1, CGI 152, KIAA1825
Rabbit
IgG
150 μL
-20°C
Primary
170759, 290673, 57130
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
ATP13A1
Liquid
Atp13a1
ATP13A1 Fusion Protein Ag9174
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat