Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP1A4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109429
Description
Immunogen sequence: MGLWGKKGTV APHDQSPRRR PKKGLIKKKM VKREKQKRNM EELKKEVVMD DHKLTLEELS TKYSVDLTKG HSHQRAKEIL TRGGPNTVTP.
This is the catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of sodium and potassium ions across the plasma membrane. This action creates the electrochemical gradient of sodium and potassium ions, providing the energy for active transport of various nutrients. Plays a role in sperm motility.
Specifications
ATP1A4 | |
Polyclonal | |
Unconjugated | |
ATP1A4 | |
3.6.3.9; ATP1A1; ATP1A4; ATP1AL2; ATPase Na+/K+ transporting subunit alpha 4; ATPase, Na+/K+ transporting, alpha 4 polypeptide; ATPase, Na+/K+ transporting, alpha polypeptide-like 2; Na(+)/K(+) ATPase alpha-4 subunit; Na,K-ATPase subunit alpha-C; Na+/K+ ATPase 4; Na+/K+ ATPase alpha 4 subunit; Na+/K+ ATPase, alpha-D polypeptide; Sodium Potassium ATPase; sodium pump 4; sodium pump subunit alpha-4; sodium/potassium-transporting ATPase alpha-4 chain; sodium/potassium-transporting ATPase subunit alpha-4; sodium-potassium ATPase catalytic subunit alpha-4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
27222, 29132 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q64541, Q9WV27 | |
ATP1A4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human ATP1A4 (NP_653300.2). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction