Learn More
ATP1B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 223381AP
Catalog No. 50-173-3836
Add to cart
Description
Description
This antibody recognizes the endogenous ATP1B2 protein in human brain. The bands between 45 kDa and 65 kDa represent the glycosylated forms of ATP1B2 in different levels.
This is the non-catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of Na+ and K+ ions across the plasma membrane. The exact function of the beta-2 subunit is not known. Mediates cell adhesion of neurons and astrocytes, and promotes neurite outgrowth.
Specifications
Specifications
ATP1B2
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13638, P14231, P14415
AMOG, ATP1B2, ATPase Na+/K+ beta 2
Rabbit
IgG
150 μL
-20°C
Primary
11932, 24214, 482
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ATP1B2
Liquid
ATP1B2
ATP1B2 Fusion Protein Ag17818
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat